Learn More
International Enviroguard™ 3 Pocket SMS Lab Coat with Knit Wrists (Blue)
SMS lab coats provide an excellent particulate barrier along with protection from light, liquid splashes and sprays. SMS fabric is strong, yet breathable, to keep users cool and help avoid heat stress. Recommended applications include pharma, labs, healthcare, controlled environments and more.
Manufacturer:  International Enviroguard™ K2228B2XL
 View more versions of this product
Catalog No. 19-472-425
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Three-layer SMS is lightweight and breathable
  • Knit wrists and collar
  • 1 left chest pocket
  • 2 hip pockets
  • Specifications
    Specifications
    Snaps
    Knit
    SMS
    2X-Large
    Blue
    General Protection
    3
    38.10 cm