Learn More
International Enviroguard™ 3 Pocket SMS Lab Coat with Knit Wrists (Blue)
SMS lab coats provide an excellent particulate barrier along with protection from light, liquid splashes and sprays. SMS fabric is strong, yet breathable, to keep users cool and help avoid heat stress. Recommended applications include pharma, labs, healthcare, controlled environments and more.
Manufacturer:  International Enviroguard™ K2228B4XL
 View more versions of this product
Catalog No. 19-472-426
Add to cart
Description
Description
  • Three-layer SMS is lightweight and breathable
  • Knit wrists and collar
  • 1 left chest pocket
  • 2 hip pockets
  • Specifications
    Specifications
    Safety
    Blue
    General Protection
    3
    40.64 cm
    Snaps
    Knit
    SMS
    4X-Large