Learn More
Eisco™ 3 Prong Double Adjustable Universal Clamp - Jumbo Sized
SCI_ED
Catalog No. s24283 Shop All Eisco Products
Click to view available options
Color:
Silver
These clamps provide a firm grip to glassware items in the labs. The jaws are adjustable and are coated with non-slip vinyl sleeve. These clamps can hold objects up to 2.28 in.. Overall length is 8 in. and the clamp works on a rod of up to .8 in..
Specifications
Type Clamp
Material Steel
Color Silver
Jaw Type 3-Prong
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.