Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Propionylpyridine, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
1570-48-5
C8H9NO
135.17
VDNKJMUNLKAGAM-UHFFFAOYSA-N
3-propionylpyridine,1-3-pyridyl propan-1-one,ethyl 3-pyridyl ketone,1-pyridin-3-yl propan-1-one,3-pyridyl ethyl ketone,1-3-pyridinyl-1-propanone,3-propionyl pyridine,1-pyridin-3-yl-propan-1-one,ethyl pyridin-3-yl ketone,1-propanone, 1-3-pyridinyl
74074
1-pyridin-3-ylpropan-1-one
CCC(=O)C1=CN=CC=C1
1570-48-5
C8H9NO
135.17
VDNKJMUNLKAGAM-UHFFFAOYSA-N
3-propionylpyridine,1-3-pyridyl propan-1-one,ethyl 3-pyridyl ketone,1-pyridin-3-yl propan-1-one,3-pyridyl ethyl ketone,1-3-pyridinyl-1-propanone,3-propionyl pyridine,1-pyridin-3-yl-propan-1-one,ethyl pyridin-3-yl ketone,1-propanone, 1-3-pyridinyl
74074
1-pyridin-3-ylpropan-1-one
CCC(=O)C1=CN=CC=C1
Infrared Spectrum Authentic
Quantity 5g
Boiling Point 106.0°C (3.0 Torr)
Assay Percent Range 97.5% min. (GC)
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material 3-Propionylpyridine

GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 216-380-8