Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridazinecarboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
2164-61-6
C5H4N2O2
124.1
RUUOPSRRIKJHNH-UHFFFAOYSA-N
3-pyridazinecarboxylic acid,3-carboxypyridazine,3-pyridazine carboxylic acid,pubchem22109,pyridazine carboxylic acid,ksc202q0p,pyridazine-3-carboxylic acid,3-carboxypyridazine; 3-pyridazinecarboxylic acid;
269369
pyridazine-3-carboxylic acid
C1=CC(=NN=C1)C(=O)O
2164-61-6
C5H4N2O2
124.1
RUUOPSRRIKJHNH-UHFFFAOYSA-N
3-pyridazinecarboxylic acid,3-carboxypyridazine,3-pyridazine carboxylic acid,pubchem22109,pyridazine carboxylic acid,ksc202q0p,pyridazine-3-carboxylic acid,3-carboxypyridazine; 3-pyridazinecarboxylic acid;
269369
pyridazine-3-carboxylic acid
C1=CC(=NN=C1)C(=O)O
Quantity 1g
Packaging Glass bottle
Melting Point 190.8°C to 194.0°C
Assay Percent Range 97%
Formula Weight 124.1
Percent Purity 97%
Chemical Name or Material 3-Pyridazinecarboxylic acid