Alfa Aesar™ 3-Pyridineacetonitrile, 98%

Manufacturer: Alfa Aesar™ B2496403

Quantity 1g

 View more versions of this product

Catalog No. AAB2496403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6443-85-2
    Molecular Formula C7H6N2
    Formula Weight 118.14
    MDL Number MFCD00006406
    UN Number UN3276
    Synonym 3-Pyridylacetonitrile
    Chemical Name or Material 3-Pyridineacetonitrile

    Specifications

    Formula Weight 118.14
    TSCA TSCA
    Molecular Formula C7H6N2
    MDL Number MFCD00006406
    CAS 6443-85-2
    UN Number UN3276
    Synonym 3-Pyridylacetonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 3-Pyridineacetonitrile
    Assay 98%

    Description

    Applications
    3-Pyridineacetonitrile can react with thiophene-2-carbaldehyde to get 2-pyridin-3-yl-3-thiophen-2-yl-acrylonitrile. It is also used as pharmaceutical intermediates.

    Solubility
    Soluble in alcohol, ether, ketone. Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at 4°C.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit