Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Pyridineacetonitrile, 98%

Click to view available options
Quantity:
1g
5g
25g
6443-85-2
C7H6N2
118.139
MFCD00006406
OIPHWUPMXHQWLR-UHFFFAOYSA-N
3-pyridineacetonitrile,3-pyridylacetonitrile,2-pyridin-3-yl acetonitrile,pyridine-3-acetonitrile,3-cyanomethylpyridine,3-cyanomethyl pyridine,pyridin-3-ylacetonitrile,2-3-pyridyl ethanenitrile,pyridin-3-yl-acetonitrile,3-pyridyl acetonitrile
80923
2-pyridin-3-ylacetonitrile
C1=CC(=CN=C1)CC#N
6443-85-2
C7H6N2
118.139
MFCD00006406
OIPHWUPMXHQWLR-UHFFFAOYSA-N
3-pyridineacetonitrile,3-pyridylacetonitrile,2-pyridin-3-yl acetonitrile,pyridine-3-acetonitrile,3-cyanomethylpyridine,3-cyanomethyl pyridine,pyridin-3-ylacetonitrile,2-3-pyridyl ethanenitrile,pyridin-3-yl-acetonitrile,3-pyridyl acetonitrile
80923
2-pyridin-3-ylacetonitrile
C1=CC(=CN=C1)CC#N
Applications
3-Pyridineacetonitrile can react with thiophene-2-carbaldehyde to get 2-pyridin-3-yl-3-thiophen-2-yl-acrylonitrile. It is also used as pharmaceutical intermediates.

Solubility
Soluble in alcohol, ether, ketone. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at 4°C.
Quantity 5g
UN Number UN3276
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 3-Pyridineacetonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

TSCA : TSCA