Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridinealdoxime, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1193-92-6
C6H6N2O
122.127
MFCD00006409
YBKOPFQCLSPTPV-YVMONPNESA-N
nicotinaldoxime,3-pyridinealdoxime,nicotinaldehyde oxime,pyridine-3-aldoxime,3-pyridinecarboxaldehyde, oxime,nicotinaldehyde, oxime,3-pyridine-aldoxime,pyridine, 3-formyl-, oxime,hydroxyimino-3-pyridylmethane,pyridinaldoxim
5371219
(NZ)-N-(pyridin-3-ylmethylidene)hydroxylamine
C1=CC(=CN=C1)C=NO
1193-92-6
C6H6N2O
122.127
MFCD00006409
YBKOPFQCLSPTPV-YVMONPNESA-N
nicotinaldoxime,3-pyridinealdoxime,nicotinaldehyde oxime,pyridine-3-aldoxime,3-pyridinecarboxaldehyde, oxime,nicotinaldehyde, oxime,3-pyridine-aldoxime,pyridine, 3-formyl-, oxime,hydroxyimino-3-pyridylmethane,pyridinaldoxim
5371219
(NZ)-N-(pyridin-3-ylmethylidene)hydroxylamine
C1=CC(=CN=C1)C=NO
Quantity 25g
Packaging Glass bottle
Melting Point 149°C to 153°C
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 122.13
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Pyridinealdoxime, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 214-782-8

RTECSNumber : UT4060000

TSCA : TSCA