Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridinecarboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
500-22-1
C6H5NO
107.11
QJZUKDFHGGYHMC-UHFFFAOYSA-N
3-pyridinecarboxaldehyde,nicotinaldehyde,3-pyridinaldehyde,pyridine-3-carboxaldehyde,3-pyridylcarboxaldehyde,3-pyridylaldehyde,nicotinealdehyde,3-formylpyridine,pyridine-3-aldehyde,nicotinic aldehyde
10371
CHEBI:28345
pyridine-3-carbaldehyde
C1=CC(=CN=C1)C=O
500-22-1
C6H5NO
107.11
QJZUKDFHGGYHMC-UHFFFAOYSA-N
3-pyridinecarboxaldehyde,nicotinaldehyde,3-pyridinaldehyde,pyridine-3-carboxaldehyde,3-pyridylcarboxaldehyde,3-pyridylaldehyde,nicotinealdehyde,3-formylpyridine,pyridine-3-aldehyde,nicotinic aldehyde
10371
CHEBI:28345
pyridine-3-carbaldehyde
C1=CC(=CN=C1)C=O
Flash Point 84°C
Specific Gravity 1.142
Color Yellow to Brown
Packaging Glass bottle
Refractive Index 1.5480 to 1.5500
Density 1.1420g/mL
Quantity 25g
Infrared Spectrum Authentic
Boiling Point 95.0°C to 97.0°C (15.0mmHg)
Assay Percent Range 98%
Linear Formula (C5H4N)COH
Beilstein 21,I,288
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 107.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Pyridinecarboxaldehyde
Show More Show Less