Alfa Aesar™ 3-Pyridinemethanol, 98%

Manufacturer: Alfa Aesar™ A1038130

Quantity 250g

 View more versions of this product

Catalog No. AAA1038130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-55-0
    Molecular Formula C6H7NO
    Formula Weight 109.13
    MDL Number MFCD00006407
    Beilstein 107851
    Merck Index 14,6527
    Synonym 3-(Hydroxymethyl)pyridine; Nicotinyl alcohol
    Chemical Name or Material 3-Pyridinemethanol

    Specifications

    Formula Weight 109.13
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Beilstein 107851
    Synonym 3-(Hydroxymethyl)pyridine; Nicotinyl alcohol
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2°C to 8°C
    Flash Point 149°C (300°F)
    Quantity 250g
    Molecular Formula C6H7NO
    CAS 100-55-0
    MDL Number MFCD00006407
    Merck Index 14,6527
    RTECS Number UT4690000
    Boiling Point 154°C (28mmHg)
    Melting Point −8° to −6°C
    Chemical Name or Material 3-Pyridinemethanol
    Assay 98%

    Description

    Solubility
    Fully miscible in water.

    Notes
    Hygroscopic. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit