Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridinepropionic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
10g
Packaging:
Glass bottle
3724-19-4
C8H9NO2
151.16
MFCD00274197
WDGXIUUWINKTGP-UHFFFAOYSA-N
3-3-pyridyl propionic acid,3-pyridinepropionic acid,3-pyridinepropanoic acid,3-pyridin-3-yl propanoic acid,3-3-pyridyl propionicacid,pyridine-3-propionic acid,3-3-pyridyl propanoic acid,3-pyridin-3-yl propionic acid,pubchem8701,maybridge4_001332
259624
3-pyridin-3-ylpropanoic acid
C1=CC(=CN=C1)CCC(=O)O
3724-19-4
C8H9NO2
151.16
MFCD00274197
WDGXIUUWINKTGP-UHFFFAOYSA-N
3-3-pyridyl propionic acid,3-pyridinepropionic acid,3-pyridinepropanoic acid,3-pyridin-3-yl propanoic acid,3-3-pyridyl propionicacid,pyridine-3-propionic acid,3-3-pyridyl propanoic acid,3-pyridin-3-yl propionic acid,pubchem8701,maybridge4_001332
259624
3-pyridin-3-ylpropanoic acid
C1=CC(=CN=C1)CCC(=O)O
Melting Point 153°C to 156°C
Color Beige
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 151.16
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Pyridinepropionic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning