Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridylacetic acid hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
2.5g
25g
Packaging:
Glass bottle
6419-36-9
C7H7NO2·ClH
173.6
MFCD00012819
XVCCOEWNFXXUEV-UHFFFAOYSA-N
3-pyridylacetic acid hydrochloride,2-pyridin-3-yl acetic acid hydrochloride,pyridine-3-acetic acid hydrochloride,3-pyridineacetic acid hydrochloride,3-pyridylacetic acid hcl,pyridin-3-ylacetic acid hydrochloride,3-pyridineacetic acid hcl,3-pyridinylacetic acid hcl,pyridine-3-acetic acid hcl,2-3-pyridinyl acetic acid hydrochloride
2723724
2-pyridin-3-ylacetic acid;hydrochloride
C1=CC(=CN=C1)CC(=O)O.Cl
6419-36-9
C7H7NO2·ClH
173.6
MFCD00012819
XVCCOEWNFXXUEV-UHFFFAOYSA-N
3-pyridylacetic acid hydrochloride,2-pyridin-3-yl acetic acid hydrochloride,pyridine-3-acetic acid hydrochloride,3-pyridineacetic acid hydrochloride,3-pyridylacetic acid hcl,pyridin-3-ylacetic acid hydrochloride,3-pyridineacetic acid hcl,3-pyridinylacetic acid hcl,pyridine-3-acetic acid hcl,2-3-pyridinyl acetic acid hydrochloride
2723724
2-pyridin-3-ylacetic acid;hydrochloride
C1=CC(=CN=C1)CC(=O)O.Cl
Melting Point 160°C to 161°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige or White to Yellow
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H4N)CH2OOH·HCl
Merck Index 10, 4635
Solubility Information Solubility in water: soluble
Formula Weight 173.6
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity ≥97.5%
Chemical Name or Material 3-Pyridylacetic acid hydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.