Alfa Aesar™ 3-Pyridyloxyacetic acid, 98%

Manufacturer: Alfa Aesar™ B2478014

Quantity 25g

 View more versions of this product

Catalog No. AAB2478014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 86649-57-2
    Molecular Formula C7H7NO3
    Formula Weight 153.14
    MDL Number MFCD04039772
    Chemical Name or Material 3-Pyridyloxyacetic acid

    Specifications

    Formula Weight 153.14
    Molecular Formula C7H7NO3
    MDL Number MFCD04039772
    CAS 86649-57-2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 210° to 212°C
    Quantity 25g
    Chemical Name or Material 3-Pyridyloxyacetic acid
    Assay 98%

    Description

    Applications
    3-Pyridyloxyacetic acid is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

    Solubility
    Slightly Soluble in water (2.0 g/L)

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit