Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Pyrroline, tech. 85% (remainder pyrrolidine)

Manufacturer:  Alfa Aesar™ H2607214

Quantity 25g

 View more versions of this product

Catalog No. AAH2607214


  Check Availability
Add to cart

Specifications

Description

Applications
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.

Solubility
Soluble in alcohol and ether. Miscible with water.

Notes
Air sensitive. Store at -20°C. Store away from oxidizing agents, air.
Description & Specifications

Chemical Identifiers

109-96-6
69.11
UN2733
14,8017
Technical Grade
C4H7N
MFCD00005213
103173
3-Pyrroline

Specifications

25g
Unpleasant
69.11
UN2733
14,8017
−18°C (0°F)
85%, (remainder pyrrolidine)
109-96-6
C4H7N
MFCD00005213
103173
90° to 91°C
3-Pyrroline
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H302
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit