Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Quinuclidinone hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
1193-65-3
C7H12ClNO
161.629
MFCD00137391
RFDPHKHXPMDJJD-UHFFFAOYSA-N
3-quinuclidinone hydrochloride,quinuclidin-3-one hydrochloride,3-quinuclidone hydrochloride,1-azabicyclo 2.2.2 octan-3-one hydrochloride,3-quinuclidnone hydrochloride,1-azabicyclo 2.2.2 octan-3-one, hydrochloride,1-aza-3-oxobicyclo 2.2.2 octane,quinuclidin-3-one, chloride,1-azabicyclo 2.2.2 octan-3-one, hydrochloride 1:1
102019
1-azabicyclo[2.2.2]octan-3-one;hydrochloride
C1CN2CCC1C(=O)C2.Cl
1193-65-3
C7H12ClNO
161.629
MFCD00137391
RFDPHKHXPMDJJD-UHFFFAOYSA-N
3-quinuclidinone hydrochloride,quinuclidin-3-one hydrochloride,3-quinuclidone hydrochloride,1-azabicyclo 2.2.2 octan-3-one hydrochloride,3-quinuclidnone hydrochloride,1-azabicyclo 2.2.2 octan-3-one, hydrochloride,1-aza-3-oxobicyclo 2.2.2 octane,quinuclidin-3-one, chloride,1-azabicyclo 2.2.2 octan-3-one, hydrochloride 1:1
102019
1-azabicyclo[2.2.2]octan-3-one;hydrochloride
C1CN2CCC1C(=O)C2.Cl
Color White
Infrared Spectrum Authentic
Melting Point >300.0°C
Loss on Drying 0.5% max. (100°C, 2 hrs)
Packaging Plastic bottle
Quantity 500g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 161.63
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3-Quinuclidinone hydrochloride, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning