Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Sulfobenzoic acid disodium salt monohydrate, 97%

Click to view available options
Quantity:
25g
100g
500g
14995-40-5
C7H6Na2O6S
264.159
MFCD00149379
VODZKGQSNPCTAD-UHFFFAOYSA-L
sodium 3-sulfonatobenzoate hydrate,3-sulfobenzoic acid disodium salt monohydrate,disodium 3-sulfonatobenzoate hydrate
2734925
disodium;3-sulfonatobenzoate;hydrate
C1=CC(=CC(=C1)S(=O)(=O)[O-])C(=O)[O-].O.[Na+].[Na+]
14995-40-5
C7H6Na2O6S
264.159
MFCD00149379
VODZKGQSNPCTAD-UHFFFAOYSA-L
sodium 3-sulfonatobenzoate hydrate,3-sulfobenzoic acid disodium salt monohydrate,disodium 3-sulfonatobenzoate hydrate
2734925
disodium;3-sulfonatobenzoate;hydrate
C1=CC(=CC(=C1)S(=O)(=O)[O-])C(=O)[O-].O.[Na+].[Na+]
Quantity 100g
Formula Weight 264.17 (246.15 Anhydrous)
Percent Purity 97%
Chemical Name or Material 3-Sulfobenzoic acid disodium salt monohydrate