Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Sulfobenzoic acid monosodium salt, 97%

Click to view available options
Quantity:
25g
100g
500g
17625-03-5
C7H5NaO5S
224.162
MFCD00066378
KQHKITXZJDOIOD-UHFFFAOYSA-M
sodium 3-sulfobenzoate,3-sulfobenzoic acid monosodium salt,benzoic acid, 3-sulfo-, monosodium salt,benzoic acid, 3-sulfo-, sodium salt 1:1,sodium hydrogen m-sulphonatobenzoate,sodium3-sulfobenzoate,sodium m-sulfobenzoate,acmc-209sh5,3-sodiosulfo benzoic acid,ksc491e3l
23668512
sodium;3-sulfobenzoate
C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]
17625-03-5
C7H5NaO5S
224.162
MFCD00066378
KQHKITXZJDOIOD-UHFFFAOYSA-M
sodium 3-sulfobenzoate,3-sulfobenzoic acid monosodium salt,benzoic acid, 3-sulfo-, monosodium salt,benzoic acid, 3-sulfo-, sodium salt 1:1,sodium hydrogen m-sulphonatobenzoate,sodium3-sulfobenzoate,sodium m-sulfobenzoate,acmc-209sh5,3-sodiosulfo benzoic acid,ksc491e3l
23668512
sodium;3-sulfobenzoate
C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]
Quantity 100g
Formula Weight 224.17
Percent Purity 97%
Chemical Name or Material 3-Sulfobenzoic acid monosodium salt

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA