Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid, 95%, Acros Organics™

Click to view available options
Quantity:
5g
50g
175277-11-9
C9H14N2O2
182.22
SFSXXMXHJOSBAZ-UHFFFAOYSA-N
3-tert-butyl-1-methyl-1h-pyrazole-5-carboxylic acid,3-tert-butyl-1-methylpyrazole-5-carboxylic acid,5-tert-butyl-2-methyl-2h-pyrazole-3-carboxylic acid,1h-pyrazole-5-carboxylic acid, 3-1,1-dimethylethyl-1-methyl,3-t-butyl-1-methyl-1h-pyrazole-5-carboxylic acid,maybridge1_008671,acmc-1bzn5,butylmethylpyrazolecarboxylicacid
720529
5-tert-butyl-2-methylpyrazole-3-carboxylic acid
CC(C)(C)C1=NN(C(=C1)C(=O)O)C
175277-11-9
C9H14N2O2
182.22
SFSXXMXHJOSBAZ-UHFFFAOYSA-N
3-tert-butyl-1-methyl-1h-pyrazole-5-carboxylic acid,3-tert-butyl-1-methylpyrazole-5-carboxylic acid,5-tert-butyl-2-methyl-2h-pyrazole-3-carboxylic acid,1h-pyrazole-5-carboxylic acid, 3-1,1-dimethylethyl-1-methyl,3-t-butyl-1-methyl-1h-pyrazole-5-carboxylic acid,maybridge1_008671,acmc-1bzn5,butylmethylpyrazolecarboxylicacid
720529
5-tert-butyl-2-methylpyrazole-3-carboxylic acid
CC(C)(C)C1=NN(C(=C1)C(=O)O)C
Quantity 50g
Melting Point 151.0°C to 154.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 182.22
Percent Purity 95%
Chemical Name or Material 3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid

GHS H Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning