Alfa Aesar™ 3-tert-Butyl-1-phenyl-2-pyrazolin-5-one, 98%

Manufacturer: Alfa Aesar™ B2193003

Quantity 1g

 View more versions of this product

Catalog No. AAB2193003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6631-89-6
    Molecular Formula C13H16N2O
    Formula Weight 216.29
    MDL Number MFCD00051762
    Beilstein 4804384
    Synonym 5-tert-Butyl-2,4-dihydro-3H-pyrazol-3-one
    Chemical Name or Material 3-tert-Butyl-1-phenyl-2-pyrazolin-5-one

    Specifications

    Formula Weight 216.29
    Molecular Formula C13H16N2O
    MDL Number MFCD00051762
    CAS 6631-89-6
    Beilstein 4804384
    Synonym 5-tert-Butyl-2,4-dihydro-3H-pyrazol-3-one
    Melting Point 108° to 113°C
    Quantity 1g
    Chemical Name or Material 3-tert-Butyl-1-phenyl-2-pyrazolin-5-one
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit