Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-tert-Butyl-1-phenyl-2-pyrazolin-5-one, 98%

Click to view available options
Quantity:
1g
5g
6631-89-6
C13H16N2O
216.284
MFCD00051762
NOSULZKBEJXMKJ-UHFFFAOYSA-N
3-tert-butyl-1-phenyl-2-pyrazolin-5-one,3h-pyrazol-3-one, 5-1,1-dimethylethyl-2,4-dihydro-2-phenyl,maybridge1_001591,2-pyrazolin-5-one, 3-tert-butyl-1-phenyl,3-tert-butyl-1-phenyl-1h-pyrazol-5 4h-one,1-phenyl-3-tert-butyl-1h-pyrazole-5 4h-one,5-tert-butyl-2-phenyl-2,4-dihydro-3h-pyrazol-3-one,5-tert-butyl-2-phenyl-2,4-dihydro-3h-pyrazol-3-one #
245718
5-tert-butyl-2-phenyl-4H-pyrazol-3-one
CC(C)(C)C1=NN(C(=O)C1)C2=CC=CC=C2
6631-89-6
C13H16N2O
216.284
MFCD00051762
NOSULZKBEJXMKJ-UHFFFAOYSA-N
3-tert-butyl-1-phenyl-2-pyrazolin-5-one,3h-pyrazol-3-one, 5-1,1-dimethylethyl-2,4-dihydro-2-phenyl,maybridge1_001591,2-pyrazolin-5-one, 3-tert-butyl-1-phenyl,3-tert-butyl-1-phenyl-1h-pyrazol-5 4h-one,1-phenyl-3-tert-butyl-1h-pyrazole-5 4h-one,5-tert-butyl-2-phenyl-2,4-dihydro-3h-pyrazol-3-one,5-tert-butyl-2-phenyl-2,4-dihydro-3h-pyrazol-3-one #
245718
5-tert-butyl-2-phenyl-4H-pyrazol-3-one
CC(C)(C)C1=NN(C(=O)C1)C2=CC=CC=C2
Quantity 5g
Melting Point 108°C to 113°C
Beilstein 4804384
Formula Weight 216.29
Percent Purity 98%
Chemical Name or Material 3-tert-Butyl-1-phenyl-2-pyrazolin-5-one