Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-tert-Butyl-2-hydroxybenzaldehyde, 96%, Thermo Scientific™

Click to view available options
Quantity:
1g
24623-65-2
C11H14O2
178.23
MFCD00778877
ROILLNJICXGZQQ-UHFFFAOYSA-N
3-tert-butyl-2-hydroxybenzaldehyde, 3-tert-butylsalicylaldehyde, 3-t-butylsalicylaldehyde, benzaldehyde, 3-1,1-dimethylethyl-2-hydroxy, 3-tert-butyl-2-hydroxy-benzaldehyde, 3-tert-butyl salicylaldehyde, acmc-1ceew, 3-tertbutylsalicylaldehyde, 2-hydroxy-3-tert.butylbenzaldehyde, 3-tert-butyl-2-oxidanyl-benzaldehyde
3678675
3-tert-butyl-2-hydroxybenzaldehyde
CC(C)(C)C1=CC=CC(C=O)=C1O
24623-65-2
C11H14O2
178.23
MFCD00778877
ROILLNJICXGZQQ-UHFFFAOYSA-N
3-tert-butyl-2-hydroxybenzaldehyde, 3-tert-butylsalicylaldehyde, 3-t-butylsalicylaldehyde, benzaldehyde, 3-1,1-dimethylethyl-2-hydroxy, 3-tert-butyl-2-hydroxy-benzaldehyde, 3-tert-butyl salicylaldehyde, acmc-1ceew, 3-tertbutylsalicylaldehyde, 2-hydroxy-3-tert.butylbenzaldehyde, 3-tert-butyl-2-oxidanyl-benzaldehyde
3678675
3-tert-butyl-2-hydroxybenzaldehyde
CC(C)(C)C1=CC=CC(C=O)=C1O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 1.0500g/mL
Boiling Point 79.0°C (0.1kPa)
Infrared Spectrum Authentic
Quantity 1g
Formula Weight 178.23
Percent Purity 96%
Chemical Name or Material 3-tert-Butyl-2-hydroxybenzaldehyde

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

Warning

RUO – Research Use Only