Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-tert-Butyl-4-hydroxyanisole, 98%, ACROS Organics™
Click to view available options
Quantity:
100g
500g
Specifications
Packaging | Glass bottle |
Flash Point | 110°C |
Quantity | 100g |
Infrared Spectrum | Authentic |
Melting Point | 58.0°C to 66.0°C |
Color | White |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97.5% min. (GC) |
Solubility Information | Solubility in water: in water:(20°C) insoluble.,in ethanol:(20°C) soluble |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 204-442-7