Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Thiopheneboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
6165-69-1
C4H5BO2S
127.96
MFCD00151851
QNMBSXGYAQZCTN-UHFFFAOYSA-N
3-thiopheneboronic acid,thiophene-3-boronic acid,3-thienylboronic acid,thiophene 3-boronic acid,thiophene-3-boronicacid,thiophen-3-yl-3-boronic acid,3-thienyl boronic acid,3-thiophene boronic acid,thiophen-3-yl-boranediol,boronic acid, 3-thienyl
581760
thiophen-3-ylboronic acid
B(C1=CSC=C1)(O)O
6165-69-1
C4H5BO2S
127.96
MFCD00151851
QNMBSXGYAQZCTN-UHFFFAOYSA-N
3-thiopheneboronic acid,thiophene-3-boronic acid,3-thienylboronic acid,thiophene 3-boronic acid,thiophene-3-boronicacid,thiophen-3-yl-3-boronic acid,3-thienyl boronic acid,3-thiophene boronic acid,thiophen-3-yl-boranediol,boronic acid, 3-thienyl
581760
thiophen-3-ylboronic acid
B(C1=CSC=C1)(O)O
Melting Point 162°C to 167°C
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Gray to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 127.96
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 3-Thiopheneboronic acid, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA