Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Thiophenecarboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
498-62-4
C5H4OS
112.15
MFCD00005466
RBIGKSZIQCTIJF-UHFFFAOYSA-N
3-thiophenecarboxaldehyde,thiophene-3-carboxaldehyde,3-formylthiophene,3-thenaldehyde,3-formyl-thiophene,3-thiophenealdehyde,thiophene-3-aldehyde,3-thiophene carboxaldehyde,3-thiophenecarbaldehyde,3-thiophenaldehyde
68135
CHEBI:87611
thiophene-3-carbaldehyde
C1=CSC=C1C=O
498-62-4
C5H4OS
112.15
MFCD00005466
RBIGKSZIQCTIJF-UHFFFAOYSA-N
3-thiophenecarboxaldehyde,thiophene-3-carboxaldehyde,3-formylthiophene,3-thenaldehyde,3-formyl-thiophene,3-thiophenealdehyde,thiophene-3-aldehyde,3-thiophene carboxaldehyde,3-thiophenecarbaldehyde,3-thiophenaldehyde
68135
CHEBI:87611
thiophene-3-carbaldehyde
C1=CSC=C1C=O
Melting Point -30.0°C
Packaging Glass bottle
Density 1.2800g/mL
Quantity 100mL
Specific Gravity 1.28
Refractive Index 1.5820 to 1.5860
Color Yellow to Brown
Infrared Spectrum Authentic
Boiling Point 194.0°C to 196.0°C
Flash Point 64°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 17,III,4497
Formula Weight 112.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Thiophenecarboxaldehyde, 98%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protectio

GHS Signal Word: Warning

EINECSNumber : 207-865-5

TSCA : TSCA

Recommended Storage : Product may darken on standing