Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Thiophenemethanol, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
71637-34-8
C5H6OS
114.17
BOWIFWCBNWWZOG-UHFFFAOYSA-N
3-thiophenemethanol,3-thienylmethanol,thiophene-3-methanol,3-thiophene methanol,3-hydroxymethylthiophene,thien-3-ylmethanol,thiophen-3-yl methanol,3-thienylmethan-1-ol,3-hydroxymethyl thiophene,3-thiophenylmethanol
123570
thiophen-3-ylmethanol
C1=CSC=C1CO
71637-34-8
C5H6OS
114.17
BOWIFWCBNWWZOG-UHFFFAOYSA-N
3-thiophenemethanol,3-thienylmethanol,thiophene-3-methanol,3-thiophene methanol,3-hydroxymethylthiophene,thien-3-ylmethanol,thiophen-3-yl methanol,3-thienylmethan-1-ol,3-hydroxymethyl thiophene,3-thiophenylmethanol
123570
thiophen-3-ylmethanol
C1=CSC=C1CO
Quantity 25g
Boiling Point 73.0°C (6.0 Torr)
Flash Point 100.5°C
Specific Gravity 1.211
Density 1.2110g/mL
Assay Percent Range 96% min. (GC)
Formula Weight 114.17
Percent Purity 97%
Chemical Name or Material 3-Thiophenemethanol

EINECSNumber : 275-741-8