Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Tolylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
17933-03-8
C7H9BO2
135.96
MFCD00040198
BJQCPCFFYBKRLM-UHFFFAOYSA-N
3-tolylboronic acid,m-tolylboronic acid,3-methylphenyl boronic acid,3-methylbenzeneboronic acid,3-methyl phenyl boronic acid,3-tolyboronic acid,3-methylphenyl boranediol,m-methylphenylboronic acid,m-tolyl boronic acid
2733950
(3-methylphenyl)boronic acid
B(C1=CC(=CC=C1)C)(O)O
17933-03-8
C7H9BO2
135.96
MFCD00040198
BJQCPCFFYBKRLM-UHFFFAOYSA-N
3-tolylboronic acid,m-tolylboronic acid,3-methylphenyl boronic acid,3-methylbenzeneboronic acid,3-methyl phenyl boronic acid,3-tolyboronic acid,3-methylphenyl boranediol,m-methylphenylboronic acid,m-tolyl boronic acid
2733950
(3-methylphenyl)boronic acid
B(C1=CC(=CC=C1)C)(O)O
Quantity 1g
Melting Point 160.0°C to 162.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H4B(OH)2
Solubility Information Solubility in water: +/- 2.5% in water.
Formula Weight 135.96
Physical Form Powder and Chunks
Percent Purity 97%
Chemical Name or Material 3-Tolylboronic acid, 97%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

TSCA : TSCA