Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Trifluoroacetyl)indole, 97%

Click to view available options
Quantity:
1g
5g
14618-45-2
C10H6F3NO
213.159
MFCD00182114
LCMDCXWSHDFQKP-UHFFFAOYSA-N
3-trifluoroacetyl indole,2,2,2-trifluoro-1-1h-indol-3-yl ethanone,2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone,3-trifluoroacetylindole,indole3trifluoroacetyl,2,2,2-trifluoro-1-1h-indol-3-yl-ethanone,3-trifluoroacetyl-1h-indole,2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one,ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl,2,2,2-trifluoro-1-indol-3-ylethan-1-one
589126
2,2,2-trifluoro-1-(1H-indol-3-yl)ethanone
C1=CC=C2C(=C1)C(=CN2)C(=O)C(F)(F)F
14618-45-2
C10H6F3NO
213.159
MFCD00182114
LCMDCXWSHDFQKP-UHFFFAOYSA-N
3-trifluoroacetyl indole,2,2,2-trifluoro-1-1h-indol-3-yl ethanone,2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone,3-trifluoroacetylindole,indole3trifluoroacetyl,2,2,2-trifluoro-1-1h-indol-3-yl-ethanone,3-trifluoroacetyl-1h-indole,2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one,ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl,2,2,2-trifluoro-1-indol-3-ylethan-1-one
589126
2,2,2-trifluoro-1-(1H-indol-3-yl)ethanone
C1=CC=C2C(=C1)C(=CN2)C(=O)C(F)(F)F
Melting Point 211°C to 214°C
Quantity 1g
Beilstein 181739
Formula Weight 213.16
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-(Trifluoroacetyl)indole