Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Trifluoromethoxy)anisole, 97+%

Click to view available options
Quantity:
1g
5g
142738-94-1
C8H7F3O2
192.137
MFCD01631504
OKSRAPDSZKICKZ-UHFFFAOYSA-N
3-trifluoromethoxy anisole,1-methoxy-3-trifluoromethoxy benzene,benzene, 1-methoxy-3-trifluoromethoxy,trifluoro 3-methoxyphenoxy methane,pubchem4158,3-trifluoromethoxyanisole,acmc-1c79q,ksc494o1j,1-methoxy-3-trifluoromethoxy-benzene,1-methoxy-3-trifluoromethyloxy benzene
2777188
1-methoxy-3-(trifluoromethoxy)benzene
COC1=CC(=CC=C1)OC(F)(F)F
142738-94-1
C8H7F3O2
192.137
MFCD01631504
OKSRAPDSZKICKZ-UHFFFAOYSA-N
3-trifluoromethoxy anisole,1-methoxy-3-trifluoromethoxy benzene,benzene, 1-methoxy-3-trifluoromethoxy,trifluoro 3-methoxyphenoxy methane,pubchem4158,3-trifluoromethoxyanisole,acmc-1c79q,ksc494o1j,1-methoxy-3-trifluoromethoxy-benzene,1-methoxy-3-trifluoromethyloxy benzene
2777188
1-methoxy-3-(trifluoromethoxy)benzene
COC1=CC(=CC=C1)OC(F)(F)F
Quantity 5g
Formula Weight 192.14
Percent Purity ≥97%
Chemical Name or Material 3-(Trifluoromethoxy)anisole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning