Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethoxy)benzenesulfonyl chloride, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
220227-84-9
C7H4ClF3O3S
260.62
DODDSXTWDSJCDN-UHFFFAOYSA-N
3-trifluoromethoxy benzenesulfonyl chloride, 3-trifluoromethoxy benzene-1-sulfonyl chloride, 3-trifluoromethoxy benzenesulphonyl chloride, 3-trifluoromethoxy-benzenesulfonyl chloride, 3-trifluoromethoxybenzenesulfonylchloride, 3-trifluoromethyl oxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3-trifluoromethoxy, pubchem5187, acmc-1ce4e
2777218
3-(trifluoromethoxy)benzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)OC(F)(F)F
220227-84-9
C7H4ClF3O3S
260.62
DODDSXTWDSJCDN-UHFFFAOYSA-N
3-trifluoromethoxy benzenesulfonyl chloride, 3-trifluoromethoxy benzene-1-sulfonyl chloride, 3-trifluoromethoxy benzenesulphonyl chloride, 3-trifluoromethoxy-benzenesulfonyl chloride, 3-trifluoromethoxybenzenesulfonylchloride, 3-trifluoromethyl oxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3-trifluoromethoxy, pubchem5187, acmc-1ce4e
2777218
3-(trifluoromethoxy)benzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)OC(F)(F)F
Infrared Spectrum Authentic
NMR Consistent with structure (1H−NMR)
Boiling Point 73.0°C to 74.0°C (0.7mmHg)
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 260.62
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethoxy)benzenesulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger

RUO – Research Use Only