Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Trifluoromethoxy)benzoic acid, 99%

Click to view available options
Quantity:
1g
5g
25g
1014-81-9
C8H5F3O3
206.12
MFCD00041501
OKPFIWIMBJNFSE-UHFFFAOYSA-N
3-trifluoromethoxy benzoic acid,m-trifluoromethoxybenzoic acid,m-trifluoromethoxy benzoic acid,benzoic acid, 3-trifluoromethoxy,3-trifluoromethoxy benzoicacid,alpha,alpha,alpha-trifluoro-m-anisic acid,3-carboxy-alpha,alpha,alpha-trifluoroanisole,3-trifluoromethoxy-benzoic acid,pubchem4590,acmc-1btyb
66096
3-(trifluoromethoxy)benzoic acid
C1=CC(=CC(=C1)OC(F)(F)F)C(=O)O
1014-81-9
C8H5F3O3
206.12
MFCD00041501
OKPFIWIMBJNFSE-UHFFFAOYSA-N
3-trifluoromethoxy benzoic acid,m-trifluoromethoxybenzoic acid,m-trifluoromethoxy benzoic acid,benzoic acid, 3-trifluoromethoxy,3-trifluoromethoxy benzoicacid,alpha,alpha,alpha-trifluoro-m-anisic acid,3-carboxy-alpha,alpha,alpha-trifluoroanisole,3-trifluoromethoxy-benzoic acid,pubchem4590,acmc-1btyb
66096
3-(trifluoromethoxy)benzoic acid
C1=CC(=CC(=C1)OC(F)(F)F)C(=O)O
Melting Point 88°C to 89°C
Quantity 1g
Beilstein 2579574
Formula Weight 206.12
Percent Purity 99%
Chemical Name or Material 3-(Trifluoromethoxy)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 213-802-2

TSCA : No

Recommended Storage : Ambient temperatures