Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Trifluoromethoxy)cinnamic acid, 96%

Click to view available options
Quantity:
0.25g
1g
168833-80-5
C10H7F3O3
232.158
MFCD00066337
CLKZZEYGXRWYNI-SNAWJCMRSA-N
3-trifluoromethoxy cinnamic acid,3-3-trifluoromethoxy phenyl acrylic acid,trans-3-trifluoromethoxy cinnamic acid,2e-3-3-trifluoromethoxy phenyl prop-2-enoic acid,rarechem bk hw 0216,3-3-trifluoromethoxy-phenyl-acrylic acid,e-3-3-trifluoromethoxy phenyl acrylic acid,e-3-3-trifluoromethoxy phenyl prop-2-enoic acid,3-3-trifluoromethoxy phenyl prop-2-enoic acid,pubchem4353
735840
(E)-3-[3-(trifluoromethoxy)phenyl]prop-2-enoic acid
C1=CC(=CC(=C1)OC(F)(F)F)C=CC(=O)O
168833-80-5
C10H7F3O3
232.158
MFCD00066337
CLKZZEYGXRWYNI-SNAWJCMRSA-N
3-trifluoromethoxy cinnamic acid,3-3-trifluoromethoxy phenyl acrylic acid,trans-3-trifluoromethoxy cinnamic acid,2e-3-3-trifluoromethoxy phenyl prop-2-enoic acid,rarechem bk hw 0216,3-3-trifluoromethoxy-phenyl-acrylic acid,e-3-3-trifluoromethoxy phenyl acrylic acid,e-3-3-trifluoromethoxy phenyl prop-2-enoic acid,3-3-trifluoromethoxy phenyl prop-2-enoic acid,pubchem4353
735840
(E)-3-[3-(trifluoromethoxy)phenyl]prop-2-enoic acid
C1=CC(=CC(=C1)OC(F)(F)F)C=CC(=O)O
Melting Point 92°C to 95°C
Quantity 0.25g
Formula Weight 232.16
Percent Purity 96%
Chemical Name or Material 3-(Trifluoromethoxy)cinnamic acid

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger