Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethy)pyridine-2-carboxylic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
87407-12-3
C7H4F3NO2
191.11
DMWLBOPLZYJGPT-UHFFFAOYSA-N
3-trifluoromethyl pyridine-2-carboxylic acid,3-trifluoromethyl picolinic acid,3-trifluoromethyl-2-pyridinecarboxylic acid,2-pyridinecarboxylic acid, 3-trifluoromethyl,3-trifluoromethyl-2-picolinic acid,acmc-209qkb,ksc447o0j,3-trifluoromethylpicolinic acid,trifluoromethyl-2-picolinic acid
13233844
3-(trifluoromethyl)pyridine-2-carboxylic acid
C1=CC(=C(N=C1)C(=O)O)C(F)(F)F
87407-12-3
C7H4F3NO2
191.11
DMWLBOPLZYJGPT-UHFFFAOYSA-N
3-trifluoromethyl pyridine-2-carboxylic acid,3-trifluoromethyl picolinic acid,3-trifluoromethyl-2-pyridinecarboxylic acid,2-pyridinecarboxylic acid, 3-trifluoromethyl,3-trifluoromethyl-2-picolinic acid,acmc-209qkb,ksc447o0j,3-trifluoromethylpicolinic acid,trifluoromethyl-2-picolinic acid
13233844
3-(trifluoromethyl)pyridine-2-carboxylic acid
C1=CC(=C(N=C1)C(=O)O)C(F)(F)F
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethy)pyridine-2-carboxylic acid

GHS Signal Word: Warning