Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Trifluoromethyl)anisole, 99%

Click to view available options
Quantity:
25g
454-90-0
C8H7F3O
176.138
MFCD00000385
XHONYVFDZSPELQ-UHFFFAOYSA-N
3-trifluoromethyl anisole,1-methoxy-3-trifluoromethyl benzene,3-methoxybenzotrifluoride,3-trifluoromethylanisole,benzene, 1-methoxy-3-trifluoromethyl,3-methoxy-1-trifluoromethyl benzene,pubchem2712,3-trifluoromethylanisol,acmc-1cuj0,m-trifluoromethyl anisole
96691
1-methoxy-3-(trifluoromethyl)benzene
COC1=CC=CC(=C1)C(F)(F)F
454-90-0
C8H7F3O
176.138
MFCD00000385
XHONYVFDZSPELQ-UHFFFAOYSA-N
3-trifluoromethyl anisole,1-methoxy-3-trifluoromethyl benzene,3-methoxybenzotrifluoride,3-trifluoromethylanisole,benzene, 1-methoxy-3-trifluoromethyl,3-methoxy-1-trifluoromethyl benzene,pubchem2712,3-trifluoromethylanisol,acmc-1cuj0,m-trifluoromethyl anisole
96691
1-methoxy-3-(trifluoromethyl)benzene
COC1=CC=CC(=C1)C(F)(F)F
Boiling Point 159°C to 160°C
Quantity 25g
Flash Point 48°C (118°F)
UN Number UN1993
Beilstein 3048340
Formula Weight 176.14
Percent Purity 99%
Chemical Name or Material 3-(Trifluoromethyl)anisole

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.