Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)anisole, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
454-90-0
C8H7F3O
176.14
MFCD00000385
XHONYVFDZSPELQ-UHFFFAOYSA-N
3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
96691
1-methoxy-3-(trifluoromethyl)benzene
COC1=CC=CC(=C1)C(F)(F)F
454-90-0
C8H7F3O
176.14
MFCD00000385
XHONYVFDZSPELQ-UHFFFAOYSA-N
3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
96691
1-methoxy-3-(trifluoromethyl)benzene
COC1=CC=CC(=C1)C(F)(F)F
Packaging Glass bottle
Flash Point 48°C
Specific Gravity 1.21
Refractive Index 1.4424 to 1.4444
Color Colorless
Infrared Spectrum Authentic
Density 1.2100g/mL
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CF3C6H4OCH3
Formula Weight 176.14
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-(Trifluoromethyl)anisole
Show More Show Less

GHS H Statement
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 207-229-7