Alfa Aesar™ 3-(Trifluoromethyl)benzaldehyde, 97%

Manufacturer: Alfa Aesar™ A1046306

Quantity 5g

 View more versions of this product

Catalog No. AAA1046306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 454-89-7
    Molecular Formula C8H5F3O
    Formula Weight 174.12
    MDL Number MFCD00003373
    Beilstein 2327537
    Synonym alpha,alpha,alpha-Trifluoro-m-tolualdehyde
    Chemical Name or Material 3- (Trifluoromethyl)benzaldehyde

    Specifications

    Formula Weight 174.12
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    TSCA TSCA
    Beilstein 2327537
    Synonym alpha,alpha,alpha-Trifluoro-m-tolualdehyde
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Flash Point 68°C (154°F)
    Odor Almond-like
    Molecular Formula C8H5F3O
    MDL Number MFCD00003373
    Boiling Point 83° to 86°C (30mmHg)
    Quantity 5g
    CAS 454-89-7
    Chemical Name or Material 3- (Trifluoromethyl)benzaldehyde
    Assay 97%

    GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit