Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)benzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
454-89-7
C8H5F3O
174.12
MFCD00003373
NMTUHPSKJJYGML-UHFFFAOYSA-N
3-trifluoromethyl benzaldehyde,m-trifluoromethylbenzaldehyde,benzaldehyde, 3-trifluoromethyl,alpha,alpha,alpha-trifluoro-m-tolualdehyde,alpha,alpha,alpha-trifluoro-3-tolualdehyde,unii-at7h4h5u3e,a,a,a-trifluoro-m-tolualdehyde,3-trifluoromethyl-benzaldehyde,mtf-bad,3-formylbenzotrifluoride
67990
3-(trifluoromethyl)benzaldehyde
C1=CC(=CC(=C1)C(F)(F)F)C=O
454-89-7
C8H5F3O
174.12
MFCD00003373
NMTUHPSKJJYGML-UHFFFAOYSA-N
3-trifluoromethyl benzaldehyde,m-trifluoromethylbenzaldehyde,benzaldehyde, 3-trifluoromethyl,alpha,alpha,alpha-trifluoro-m-tolualdehyde,alpha,alpha,alpha-trifluoro-3-tolualdehyde,unii-at7h4h5u3e,a,a,a-trifluoro-m-tolualdehyde,3-trifluoromethyl-benzaldehyde,mtf-bad,3-formylbenzotrifluoride
67990
3-(trifluoromethyl)benzaldehyde
C1=CC(=CC(=C1)C(F)(F)F)C=O
Specific Gravity 1.3
Boiling Point 83°C to 86°C (30.0mmHg)
Flash Point 68°C
Density 1.3000g/mL
Refractive Index 1.4639 to 1.4659
Packaging Glass bottle
Color Colorless to Orange
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CF3C6H4CHO
Beilstein 07, III, 1016
Formula Weight 174.12
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzaldehyde, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-228-1

TSCA : TSCA