Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)benzaldehyde 95.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
454-89-7
C8H5F3O
174.122
MFCD00003373
NMTUHPSKJJYGML-UHFFFAOYSA-N
3-trifluoromethyl benzaldehyde,m-trifluoromethylbenzaldehyde,benzaldehyde, 3-trifluoromethyl,alpha,alpha,alpha-trifluoro-m-tolualdehyde,alpha,alpha,alpha-trifluoro-3-tolualdehyde,unii-at7h4h5u3e,a,a,a-trifluoro-m-tolualdehyde,3-trifluoromethyl-benzaldehyde,mtf-bad,3-formylbenzotrifluoride
67990
3-(trifluoromethyl)benzaldehyde
C1=CC(=CC(=C1)C(F)(F)F)C=O
454-89-7
C8H5F3O
174.122
MFCD00003373
NMTUHPSKJJYGML-UHFFFAOYSA-N
3-trifluoromethyl benzaldehyde,m-trifluoromethylbenzaldehyde,benzaldehyde, 3-trifluoromethyl,alpha,alpha,alpha-trifluoro-m-tolualdehyde,alpha,alpha,alpha-trifluoro-3-tolualdehyde,unii-at7h4h5u3e,a,a,a-trifluoro-m-tolualdehyde,3-trifluoromethyl-benzaldehyde,mtf-bad,3-formylbenzotrifluoride
67990
3-(trifluoromethyl)benzaldehyde
C1=CC(=CC(=C1)C(F)(F)F)C=O
Boiling Point 66°C
Color Yellow
Quantity 25g
Formula Weight 174.12
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material 3-(Trifluoromethyl)benzaldehyde

EINECSNumber : (9)-0923

TSCA : Yes