Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Trifluoromethyl)benzoylacetonitrile, 97%

Click to view available options
Quantity:
1g
5g
25g
27328-86-5
C10H6F3NO
213.159
MFCD00052315
GEPORLBYZLQDOB-UHFFFAOYSA-N
3-trifluoromethylbenzoylacetonitrile,3-oxo-3-3-trifluoromethyl phenyl propanenitrile,3-trifluoromethyl benzoylacetonitrile,2-cyano-3'-trifluoromethyl acetophenone,3-oxo-3-3-trifluoromethyl-phenyl-propionitrile,pubchem12076,acmc-1cbhg,maybridge1_000123,3-trifluoromethyl benzoyl acetonitrile
520304
3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile
C1=CC(=CC(=C1)C(F)(F)F)C(=O)CC#N
27328-86-5
C10H6F3NO
213.159
MFCD00052315
GEPORLBYZLQDOB-UHFFFAOYSA-N
3-trifluoromethylbenzoylacetonitrile,3-oxo-3-3-trifluoromethyl phenyl propanenitrile,3-trifluoromethyl benzoylacetonitrile,2-cyano-3'-trifluoromethyl acetophenone,3-oxo-3-3-trifluoromethyl-phenyl-propionitrile,pubchem12076,acmc-1cbhg,maybridge1_000123,3-trifluoromethyl benzoyl acetonitrile
520304
3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile
C1=CC(=CC(=C1)C(F)(F)F)C(=O)CC#N
Applications
3-(Trifluoromethyl)benzoylacetonitrile is used as a pharmaceutical intermediate.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Melting Point 58°C to 63°C
Quantity 25g
Beilstein 2373428
Formula Weight 213.16
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzoylacetonitrile

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures