Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)benzyl alcohol, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
349-75-7
C8H7F3O
176.138
MFCD00004645
BXEHKCUWIODEDE-UHFFFAOYSA-N
3-trifluoromethyl benzyl alcohol,3-trifluoromethylbenzyl alcohol,3-trifluoromethyl phenyl methanol,3-trifluoromethyl benzylalcohol,m-trifluoromethylbenzyl alcohol,m-trifluoromethyl benzyl alcohol,3-trifluoromethyl benzylic alcohol,benzenemethanol, 3-trifluoromethyl,m-trifluorome benzyl alcohol
67681
[3-(trifluoromethyl)phenyl]methanol
C1=CC(=CC(=C1)C(F)(F)F)CO
349-75-7
C8H7F3O
176.138
MFCD00004645
BXEHKCUWIODEDE-UHFFFAOYSA-N
3-trifluoromethyl benzyl alcohol,3-trifluoromethylbenzyl alcohol,3-trifluoromethyl phenyl methanol,3-trifluoromethyl benzylalcohol,m-trifluoromethylbenzyl alcohol,m-trifluoromethyl benzyl alcohol,3-trifluoromethyl benzylic alcohol,benzenemethanol, 3-trifluoromethyl,m-trifluorome benzyl alcohol
67681
[3-(trifluoromethyl)phenyl]methanol
C1=CC(=CC(=C1)C(F)(F)F)CO
Packaging Glass bottle
Quantity 5g
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Color Colorless to Yellow
Density 1.2900g/mL
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula CF3C6H4CH2OH
Formula Weight 176.14
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material 3-(Trifluoromethyl)benzyl alcohol, 98+%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 206-489-9

TSCA : TSCA