Alfa Aesar™ 3-(Trifluoromethyl)benzylamine, 97%

Manufacturer: Alfa Aesar™ B2414306

Quantity 5g

 View more versions of this product

Catalog No. AAB2414306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2740-83-2
    Molecular Formula C8H8F3N
    Formula Weight 175.15
    MDL Number MFCD00008117
    UN Number UN2735
    Beilstein 1102465
    Chemical Name or Material 3-(Trifluoromethyl)benzylamine

    Specifications

    Formula Weight 175.15
    CAS 2740-83-2
    UN Number UN2735
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Flash Point 71°C (159°F)
    Quantity 5g
    Molecular Formula C8H8F3N
    MDL Number MFCD00008117
    Beilstein 1102465
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Boiling Point 93° to 97°C (28mmHg)
    Chemical Name or Material 3-(Trifluoromethyl)benzylamine
    Assay 97%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit