Alfa Aesar™ 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride, 96%

Manufacturer: Alfa Aesar™ H3423503

Quantity 1g

 View more versions of this product

Catalog No. AAH3423503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 885267-96-9
    Molecular Formula C13H8ClF3O2S
    Formula Weight 320.71
    MDL Number MFCD06409268
    UN Number UN3261
    Chemical Name or Material 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride

    Specifications

    Formula Weight 320.71
    UN Number UN3261
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 78° to 92°C
    Quantity 1g
    Molecular Formula C13H8ClF3O2S
    MDL Number MFCD06409268
    CAS 885267-96-9
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride
    Assay 96%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit