Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
885267-96-9
C13H8ClF3O2S
320.72
WJSSWLPZDVPLIK-UHFFFAOYSA-N
3'-trifluoromethyl biphenyl-4-sulfonyl chloride,4-3-trifluoromethyl phenyl benzenesulfonyl chloride,3'-trifluoromethyl-1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 3'-trifluoromethyl,3'-trifluoromethyl 1,1'-biphenyl-4-sulfonyl chloride,3'-trifluoromethylbiphenyl-4-sulfonyl chloride
16244690
4-[3-(trifluoromethyl)phenyl]benzenesulfonyl chloride
C1=CC(=CC(=C1)C(F)(F)F)C2=CC=C(C=C2)S(=O)(=O)Cl
885267-96-9
C13H8ClF3O2S
320.72
WJSSWLPZDVPLIK-UHFFFAOYSA-N
3'-trifluoromethyl biphenyl-4-sulfonyl chloride,4-3-trifluoromethyl phenyl benzenesulfonyl chloride,3'-trifluoromethyl-1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 3'-trifluoromethyl,3'-trifluoromethyl 1,1'-biphenyl-4-sulfonyl chloride,3'-trifluoromethylbiphenyl-4-sulfonyl chloride
16244690
4-[3-(trifluoromethyl)phenyl]benzenesulfonyl chloride
C1=CC(=CC(=C1)C(F)(F)F)C2=CC=C(C=C2)S(=O)(=O)Cl
Melting Point 78°C to 82°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. diethylsulfonamide derivative (HPLC)
Formula Weight 320.72
Percent Purity 97%
Chemical Name or Material 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger