Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)phenylacetone, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
2.5g
10g
Packaging:
Glass bottle
21906-39-8
C10H9F3O
202.176
MFCD00000397
JPHQCDCEBDRIOL-UHFFFAOYSA-N
3-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl propan-2-one, m-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl acetone, 3-trifluormethyl phenylacetone, 1-3-trifluoromethyl phenyl-2-propanone, m-trifluoromethylphenylacetone, 2-propanone, 1-3-trifluoromethyl phenyl
89101
1-[3-(trifluoromethyl)phenyl]propan-2-one
CC(=O)CC1=CC(=CC=C1)C(F)(F)F
21906-39-8
C10H9F3O
202.176
MFCD00000397
JPHQCDCEBDRIOL-UHFFFAOYSA-N
3-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl propan-2-one, m-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl acetone, 3-trifluormethyl phenylacetone, 1-3-trifluoromethyl phenyl-2-propanone, m-trifluoromethylphenylacetone, 2-propanone, 1-3-trifluoromethyl phenyl
89101
1-[3-(trifluoromethyl)phenyl]propan-2-one
CC(=O)CC1=CC(=CC=C1)C(F)(F)F
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.204g/mL
Refractive Index 1.4560 to 1.4580 (20°C, 589nm)
Quantity 10g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CF3C6H4CH2COCH3
Formula Weight 202.18
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.