Alfa Aesar™ 3-(Trifluoromethyl)phenylacetone, 97%, may cont. up to ca 5% monohydrate

Manufacturer: Alfa Aesar™ H2709506

Quantity 5g

 View more versions of this product

Catalog No. AAH2709506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21906-39-8
    Molecular Formula C10H9F3O
    Formula Weight 202.18
    MDL Number MFCD00000397
    Synonym 1-(alpha,alpha,alpha-Trifluoro-m-tolyl)-2-propanone
    Chemical Name or Material 3-(Trifluoromethyl)phenylacetone

    Specifications

    Formula Weight 202.18
    Molecular Formula C10H9F3O
    MDL Number MFCD00000397
    CAS 21906-39-8
    Synonym 1-(alpha,alpha,alpha-Trifluoro-m-tolyl)-2-propanone
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point >110°C (230°F)
    Quantity 5g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 89° to 90°C (0.5mmHg)
    Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
    Assay 97%, may contain up to ≈5% monohydrate

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit