Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)salicylaldehyde, 98+%, Thermo Scientific Chemicals

Catalog Number AAH32858MD
Click to view available options
Quantity:
250 mg
1 g
336628-67-2
C8H5F3O2
190.12
MFCD03788529
ZUNOXMJBNMKYOM-UHFFFAOYSA-N
2-hydroxy-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl salicylaldehyde, 2-hydroxy-3-trifluoromethyl-benzaldehyde, benzaldehyde, 2-hydroxy-3-trifluoromethyl, pubchem10111, 2-oxidanyl-3-trifluoromethyl benzaldehyde
2779274
2-hydroxy-3-(trifluoromethyl)benzaldehyde
OC1=C(C=O)C=CC=C1C(F)(F)F
336628-67-2
C8H5F3O2
190.12
MFCD03788529
ZUNOXMJBNMKYOM-UHFFFAOYSA-N
2-hydroxy-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl salicylaldehyde, 2-hydroxy-3-trifluoromethyl-benzaldehyde, benzaldehyde, 2-hydroxy-3-trifluoromethyl, pubchem10111, 2-oxidanyl-3-trifluoromethyl benzaldehyde
2779274
2-hydroxy-3-(trifluoromethyl)benzaldehyde
OC1=C(C=O)C=CC=C1C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY
Melting Point 54°C to 59°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 190.12
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)salicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit