Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trimethylsilyl)-1-propanol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
2917-47-7
C6H16OSi
132.278
MFCD00009712
YNUUCDHAZGAVEZ-UHFFFAOYSA-N
3-trimethylsilyl-1-propanol,1-propanol, 3-trimethylsilyl,3-trimethylsilyl propan-1-ol,3-trimethylsilylpropanol,3-trimethylsilyl propanol,acmc-20anhm,4,4-dimethyl-4-silapentanol,trimethyl-3-hydroxypropylsilane,1-propanol,3-trimethylsilyl
76213
3-trimethylsilylpropan-1-ol
C[Si](C)(C)CCCO
2917-47-7
C6H16OSi
132.278
MFCD00009712
YNUUCDHAZGAVEZ-UHFFFAOYSA-N
3-trimethylsilyl-1-propanol,1-propanol, 3-trimethylsilyl,3-trimethylsilyl propan-1-ol,3-trimethylsilylpropanol,3-trimethylsilyl propanol,acmc-20anhm,4,4-dimethyl-4-silapentanol,trimethyl-3-hydroxypropylsilane,1-propanol,3-trimethylsilyl
76213
3-trimethylsilylpropan-1-ol
C[Si](C)(C)CCCO
Color Colorless
Density 0.832g/mL
Packaging Glass bottle
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)3Si(CH2)3OH
Beilstein 04, III, 1854
Formula Weight 132.28
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-(Trimethylsilyl)-1-propanol, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning