Alfa Aesar™ 3-Trimethylsilyl-2-oxazolidinone

Manufacturer: Alfa Aesar™ 4120906

Quantity 5g

 View more versions of this product

Catalog No. AA4120906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 43112-38-5
    Formula Weight 159.26
    MDL Number MFCD00010197
    Chemical Name or Material 3-Trimethylsilyl-2-oxazolidinone

    Specifications

    Formula Weight 159.26
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Refractive Index 1.4540
    Physical Form Liquid
    MDL Number MFCD00010197
    Health Hazard 1 Warning
    Density 1.013g/mL
    CAS 43112-38-5
    Sensitivity Moisture Sensitive
    Boiling Point 99° to 100°C (6mmHg)
    Quantity 5g
    Flash Point 92° (198°F)
    Chemical Name or Material 3-Trimethylsilyl-2-oxazolidinone

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit