Alfa Aesar™ 3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%

Manufacturer: Alfa Aesar™ H5339714

Quantity 25g

 View more versions of this product

Catalog No. AAH5339714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87219-80-5
    Molecular Formula C10H20O2Si
    Linear Formula (CH3)3SiC≡CCH(OCH2CH3)2
    Formula Weight 200.35
    MDL Number MFCD08275372
    UN Number UN1993
    Synonym 3-(Trimethylsilyl)propargyl aldehyde diethyl acetal
    Chemical Name or Material 3-(Trimethylsilyl)propiolaldehyde diethyl acetal

    Specifications

    Formula Weight 200.35
    Linear Formula (CH3)3SiC≡CCH(OCH2CH3)2
    MDL Number MFCD08275372
    Synonym 3-(Trimethylsilyl)propargyl aldehyde diethyl acetal
    Boiling Point 170° to 172°C
    Quantity 25g
    Flash Point 40°C (104°F)
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C10H20O2Si
    CAS 87219-80-5
    UN Number UN1993
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 3-(Trimethylsilyl)propiolaldehyde diethyl acetal
    Assay 97%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit