Alfa Aesar™ 3-(Trimethylsilyl)propiolic acid, 97%

Manufacturer: Alfa Aesar™ H5338014

Quantity 25g

 View more versions of this product

Catalog No. AAH5338014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5683-31-8
    Molecular Formula C6H10O2Si
    Linear Formula (CH3)3SiC≡CCO2H
    Formula Weight 142.23
    MDL Number MFCD00190216
    Beilstein 1755674
    Chemical Name or Material 3-(Trimethylsilyl)propiolic acid

    Specifications

    Formula Weight 142.23
    Molecular Formula C6H10O2Si
    Linear Formula (CH3)3SiC≡CCO2H
    MDL Number MFCD00190216
    Melting Point 47° to 49°C
    Quantity 25g
    Flash Point 96°C (205°F)
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 5683-31-8
    Beilstein 1755674
    Boiling Point 105° to 110°C (10mmHg)
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 3-(Trimethylsilyl)propiolic acid
    Assay 97%

    Description

    Applications
    3-(Trimethylsilyl)propiolic acid may be used in the regioselective preparation of 1,5-trisubstituted 1H-1,2,3-triazoles. It is also used as pharmaceutical intermediate.

    Solubility
    Soluble in methanol.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit