Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Trimethylsilylmethyl-N-tert-butylcrotonaldimine, Acros Organics

Click to view available options
Quantity:
500mg
56637-75-3
C12H25NSi
211.424
MFCD01632576
BOLUBIYKJJKYQE-JDYWATNUSA-N
e-tert-butyl 2e-3-methyl-4-trimethylsilyl but-2-en-1-ylidene amine,3-trimethylsilylmethyl-n-tert-butylcrotonaldimine,e-n-tert-butyl-3-methyl-4-trimethylsilylbut-2-en-1-imine
5702563
(E)-N-tert-butyl-3-methyl-4-trimethylsilylbut-2-en-1-imine
CC(=CC=NC(C)(C)C)C[Si](C)(C)C
56637-75-3
C12H25NSi
211.424
MFCD01632576
BOLUBIYKJJKYQE-JDYWATNUSA-N
e-tert-butyl 2e-3-methyl-4-trimethylsilyl but-2-en-1-ylidene amine,3-trimethylsilylmethyl-n-tert-butylcrotonaldimine,e-n-tert-butyl-3-methyl-4-trimethylsilylbut-2-en-1-imine
5702563
(E)-N-tert-butyl-3-methyl-4-trimethylsilylbut-2-en-1-imine
CC(=CC=NC(C)(C)C)C[Si](C)(C)C
Color Yellow
Quantity 500mg
Infrared Spectrum Authentic
Refractive Index 1.4750 to 1.4770 (20°C, 589nm)
NMR Complies
Formula Weight 211.42
Physical Form Liquid
Chemical Name or Material 3-Trimethylsilylmethyl-N-tert-butylcrotonaldimine